Tungstosilicic acid hydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silicotungstic acid
039651-09
039651-18
AA39651-09EA
40.1
USD
AA39651-09
AA39651-18
Tungstosilicic acid hydrate, Reagent Grade
Tungstosilicic acid Hydrate
Catalyst for organic synthesis, minerals separation, reagent for alkaloids
Soluble in water
CAS No.: 12027-43-9
Molecular Formula: H4SiO4.12WO3.xH2O
Formula Weight: 2878.28(anhy)
Physical Form: Crystalline
Solubility: Soluble in water
MDL No.: MFCD00036271
Applications: Catalyst for organic synthesis, minerals separation, reagent for alkaloids
Formula:
H₆O₄₁SiW₁₂ MW: 2896.172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036271 CAS Number: 12027-43-9 Merck Index: 14,8502 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...