Tin(II) tartrate hydrate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

039604-14 039604-22
AA39604-14EA 132 USD
AA39604-14 AA39604-22
Tin(II) tartrate hydrate ≥95%
Tin(II) tartrate hydrate

Purity: 95% CAS No.: 815-85-0 Molecular Formula: C4H4O6Sn.xH2O Formula Weight: 266.76(anhy) Physical Form: Powder Sensitivity: Air Sensitive Odor: Odorless MDL No.: MFCD00054358

Formula: C₄H₄O₆Sn·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00054358
CAS Number: 815-85-0
EINECS: 212-427-1
Merck Index: 14,8792

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR