cis-Dichlorobis(triethylphosphine)platinum(II)
Supplier: Thermo Scientific Chemicals
|
Danger
|
039284-ME
AA39284-MEEA
344
USD
AA39284-ME
cis-Dichlorobis(triethylphosphine)platinum(II)
cis-Dichlorobis(triethylphosphine)platinum(II)
CAS No.: 15692-07-6
Molecular Formula: PtCl2[P(C2H5)3]2
Formula Weight: 502.32
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: 139-142°
MDL No.: MFCD00058821
Formula:
PtCl₂[P(C₂H₅)₃]₂ MW: 502.304 g/mol Melting Pt: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058821 CAS Number: 15692-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...