Aluminium perchlorate nonahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum perchlorate nonahydrate
011659-22
011659-36
AA11659-22EA
60.9
USD
AA11659-22
AA11659-36
Aluminium perchlorate nonahydrate, Reagent Grade
Aluminium perchlorate nonahydrate
CAS No.: 81029-06-3
Molecular Formula: Al(ClO4)3.9H2O
Formula Weight: 487.46 (325.33anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 82°
UN No.: 1481
MDL No.: MFCD00149130
Formula:
AlCl₃O₁₂·9H₂O Melting Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149130 CAS Number: 81029-06-3 EINECS: 238-437-6 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...