Octylphosphonic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

L18586-03 L18586-06
AAAL18586-03EA 74.1 USD
AAAL18586-03 AAAL18586-06
Octylphosphonic acid 99%
Octylphosphonic acid

Purity: 99% CAS No.: 4724-48-5 Molecular Formula: C8H19O3P Formula Weight: 194.21 Melting Point: 100-102° Beilstein Reference: 1764242

Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00015841
CAS Number: 4724-48-5
EINECS: 225-218-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR