Cyclopentyl methyl ether (CPME) ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27349-0F
H27349-AP
AAH27349-0FEA
320.38
USD
AAH27349-0F
AAH27349-AP
Cyclopentyl methyl ether (CPME) ≥99.9%
Cyclopentyl methyl ether (CPME)
Purity: 99.9+%
CAS No.: 5614-37-9
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: -1°(-30°F)
Refractive Index: 1.4210
Melting Point: -140°
Boiling Point: 106°
UN No.: 3271
MDL No.: MFCD08276401
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 106 °C Melting Pt: –140 °C Density: 0.86 Flash Pt: -1 °C(-30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08276401 CAS Number: 5614-37-9 UN: 3271 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...