4-Bromo-o-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Bromo-3,4-diaminobenzene, 4-Bromobenzene-1,2-diamine, 4-Bromo-1,2-phenylenediamine, 4-Bromo-1,2-diaminobenzene, 2-Amino-4-bromophenylamine
H26929-06
H26929-14
H26929-22
AAH26929-06EA
60.1
USD
AAH26929-06
AAH26929-14
AAH26929-22
4-Bromo-o-phenylenediamine 97%
4-Bromo-o-phenylenediamine
Purity: 97%
CAS No.: 1575-37-7
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Melting Point: 64-66°
UN No.: 2811
MDL No.: MFCD02660622
Formula:
C₆H₇BrN₂ Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD02660622 CAS Number: 1575-37-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...