Orotic acid, anhydrous 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25349-22
B25349-36
AAB25349-22EA
108
USD
AAB25349-22
AAB25349-36
Orotic acid, anhydrous 97%
Orotic acid
Purity: 97%
CAS No.: 65-86-1
Molecular Formula: C5H4N2O4
Formula Weight: 156.10
Melting Point: 344-347°
MDL No.: MFCD00006027
Formula:
C₅H₄N₂O₄ Melting Pt: 344…347 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006027 CAS Number: 65-86-1 EINECS: 200-619-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...