Isoamyl acetate 99% cont. ca 10% other isomers
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isopentyl acetate, Acetic acid isoamyl ester
B21618-AP
B21618-AE
AAAB21618-APEA
94.46
USD
AAAB21618-AP
AAAB21618-AE
Isoamyl acetate 99% cont. ca 10% other isomers
Isoamyl acetate
Purity: 99%
CAS No.: 123-92-2
Molecular Formula: CH3CO2CH2CH2CH(CH3)2
Formula Weight: 130.19
Flash Point: 25°(77°F)
Refractive Index: 1.400
Odor: Fruit-like
Melting Point: -78°
Boiling Point: 142°
Merck Reference: 14,5111
UN No.: 1104
MDL No.: MFCD00008946
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 142 °C Melting Pt: –78 °C Density: 0.876 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008946 CAS Number: 123-92-2 EINECS: 204-662-3 UN: 1104 ADR: 3,III Merck Index: 14,5111 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...