2-Oxazolidinone 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Oxazolidone, Oxazolidin-2-one
A14543-36
A14543-14
A14543-22
AAA14543-36EA
446
USD
AAA14543-36
AAA14543-14
AAA14543-22
2-Oxazolidinone 99%
2-Oxazolidinone
Fieser: 12,359
Purity: 99%
CAS No.: 497-25-6
Molecular Formula: C3H5NO2
Formula Weight: 87.08
Melting Point: 84-89°
Boiling Point: 220°/48mm
MDL No.: MFCD00005268
Formula:
C₃H₅NO₂ Boiling Pt: 220 °C/48 mm Melting Pt: 84…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005268 CAS Number: 497-25-6 EINECS: 207-840-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...