Diammonium thiosulfate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Ammonium hyposulphite, Ammonium hyposulfite, Ammonium thiosulphate, Ammonium thiosulfate

A10190-22 A10190-36 A10190-0E
AAAA10190-22EA 35.21 USD
AAAA10190-22 AAA10190-36 AAAA10190-0E
Diammonium thiosulfate 98+%
di-Ammonium thiosulphate

Purity: 98+% CAS No.: 7783-18-8 Molecular Formula: (NH4)2S2O3 Formula Weight: 148.20 pH: 6.0-6.5 Melting Point: ca 180° dec. MDL No.: MFCD00015965

Formula: H₈N₂O₃S₂
MW: 148.21 g/mol
Melting Pt: ca 180 °C (dec.)
Density: 1.64
Storage Temperature: Ambient
MDL Number: MFCD00015965
CAS Number: 7783-18-8
EINECS: 231-982-0
Merck Index: 14,562

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR