Charcoal activated -20+40 mesh

Supplier: Thermo Scientific Chemicals

Synonyms: Activated carbonCarbon activatedActivated charcoal

45478-A3 45478-36 45478-36 45478-22
AA45478-A3EA 168.88 USD
CAAA45478-36 AA45478-A3 AA45478-22 AA45478-36
Charcoal activated -20+40 mesh
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR