Arsenic(III) iodide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
045067-09
045067-18
AA45067-09EA
155
USD
AA45067-09
AA45067-18
Arsenic(III) iodide 98%
Arsenic (III) iodide
Purity: 98%
CAS No.: 7784-45-4
Molecular Formula: AsI3
Formula Weight: 455.64
Physical Form: Small Pieces
Density: 4.39
Melting Point: 140°
Boiling Point: 424°
Merck Reference: 14,803
UN No.: 1557
MDL No.: MFCD00014170
Formula:
AsI₃ Boiling Pt: 424 °C Melting Pt: 140 °C Density: 4.39 Storage Temperature: Ambient |
MDL Number:
MFCD00014170 CAS Number: 7784-45-4 EINECS: 232-068-4 UN: 1557 ADR: 6.1,II Merck Index: 14,803 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...