Carbon disulfide ≥99.9% low in benzene
Supplier: Thermo Scientific Chemicals
|
Danger
|
040910-AE
040910-AY
040910-AP
AA40910-AEEA
59.17
USD
AA40910-AE
AA40910-AY
AA40910-AP
Carbon disulfide ≥99.9% low in benzene
Carbon disulphide
1ppm max trace organic impurities (as benzene)
Purity: 99.9%
CAS No.: 75-15-0
Molecular Formula: CS2
Formula Weight: 76.14
Flash Point: -30°(-22°F)
Physical Form: Liquid
Density: 1.2632
Refractive Index: 1.6270
Odor: Unpleasant
Melting Point: -112°
Boiling Point: 46°
Merck Reference: 14,1811
UN No.: 1131
MDL No.: MFCD00011321
Formula:
CS₂ MW: 76.14 g/mol Boiling Pt: 46 °C Melting Pt: –112 °C Density: 1.2632 Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3,I Merck Index: 14,1811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...