Octylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

020651-03 020651-06
AA20651-03EA 67.4 USD
AA20651-03 AA20651-06
Octylphosphonic acid 98%
Octylphosphonic acid

Purity: 98% CAS No.: 4724-48-5 Molecular Formula: C8H19O3P Formula Weight: 194.21 Physical Form: Crystalline Melting Point: 100-102° Beilstein Reference: 1764242 MDL No.: MFCD00015841

Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00015841
CAS Number: 4724-48-5
EINECS: 225-218-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR