Octadecylphosphonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
n-Octadecylphosphonic acid, ODPA
020645-06
020645-03
AA20645-06EA
455
USD
AA20645-06
AA20645-03
Octadecylphosphonic acid 97%
Octadecylphosphonic acid
Purity: 97%
CAS No.: 4724-47-4
Molecular Formula: C18H39O3P
Formula Weight: 334.48
Physical Form: Crystalline
Appearance: White to off-white
Melting Point: 92-98°
MDL No.: MFCD00015838
Formula:
C₁₈H₃₉O₃P MW: 334.47 g/mol Melting Pt: 92…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015838 CAS Number: 4724-47-4 EINECS: 225-216-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...