Anti-IL25 Mouse Monoclonal Antibody [clone: 11K8B1]

Supplier: Rockland Immunochemical
209-301-C58 209-301-C58S
RL209-301-C58EA 565.22 USD
RL209-301-C58 RL209-301-C58S
Anti-IL25 Mouse Monoclonal Antibody [clone: 11K8B1]
Antibodies
IL 17E antibody has been tested for use in Flow Cytometry and western blotting. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000-1:50000; Flow Cytometry: 1µg/ml; Western Blot: 0.5µg/ml;

Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 11K8B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR