Anti-HAUS8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-C53 600-401-C53S
RL600-401-C53EA 458.66 USD
RL600-401-C53 RL600-401-C53S
Anti-HAUS8 Rabbit Polyclonal Antibody
Antibodies
Hice1 antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 44.9 kDa in size corresponding to human Hice1 Protein by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:30,000-1:45000; Western Blot: 0.5 µg/ml;

Type: Primary
Antigen: Hice1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR