Anti-IgG Rabbit Polyclonal Antibody (ATTO 647N)

Supplier: Rockland Immunochemical
605-456-013 605-456-013S
RL605-456-013EA 426.27 USD
RL605-456-013 RL605-456-013S
Anti-IgG Rabbit Polyclonal Antibody (ATTO 647N)
Antibodies
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 647N
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR