Anti-Annexin V Antibody (Alexa Fluor® 647)

Supplier: Biolegend
640911 640912
640911-BLEA 122.7 USD
640911-BL 640912-BL
Anti-Annexin V Antibody (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Annexin V Reactivity: All mammalian species; Apps: FC; Size: 100 tests

Each lot of this product is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per 100,000 - million cells in a 100 μl volume of Annexin V Binding Buffer (Cat No. 422201). It is recommended that the reagent be titrated for optimal performance for each application.
* Alexa Fluor® 647 has a maximum emission of 668 nm when it is excited at 633nm / 635nm.

Type: Primary
Antigen: Annexin V
Clonality:
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host:
Isotype:
Reactivity: All mammalian species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR