Anti-POLR2A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-49546SS NBP1-49546
102179-212EA 208 USD
102179-212 102179-210
Anti-POLR2A Rabbit Polyclonal Antibody
Antibodies
The RNA Polymerase II / POLR2A [p Thr4] Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNA Polymerase II / POLR2A. This antibody reacts with human, chicken, drosophila, yeast. The RNA Polymerase II / POLR2A [p Thr4] Antibody has been validated for the following applications: Western Blot, Dot Blot.

Type: Primary
Antigen: POLR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken, Drosophila, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR