Anti-PLIN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-40877 NB110-40877SS
102122-178EA 577.23 USD
102122-178 102122-192
Anti-PLIN2 Rabbit Polyclonal Antibody
Antibodies
The Perilipin-2 / ADFP Antibody from Novus Biologicals is a rabbit polyclonal antibody to Perilipin-2 / ADFP. This antibody reacts with human, mouse. The Perilipin-2 / ADFP Antibody has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR