Anti-POLR2A Mouse Monoclonal Antibody [clone: 4H8]

Supplier: Novus Biologicals
NB200-598SS NB200-598
102129-556EA 208 USD
102129-556 102129-554
Anti-POLR2A Mouse Monoclonal Antibody [clone: 4H8]
Antibodies
The RNA Polymerase II / POLR2A Antibody (4H8) from Novus Biologicals is a mouse monoclonal antibody to RNA Polymerase II / POLR2A. This antibody reacts with human, mouse, yeast, primate (negative). The RNA Polymerase II / POLR2A Antibody (4H8) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF-ready.

Type: Primary
Antigen: POLR2A
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Yeast, Primate (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR