Anti-GLO1 Mouse Monoclonal Antibody [clone: Glo1a]

Supplier: Novus Biologicals
NBP1-19015SS NBP1-19015
102158-406EA 208 USD
102158-406 102158-394
Anti-GLO1 Mouse Monoclonal Antibody [clone: Glo1a]
Antibodies
Mouse Monoclonal GLO1 Antibody (Glo1a). Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: GLO1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR