Anti-DNA Ligase III Mouse Monoclonal Antibody [clone: 1F3]

Supplier: Novus Biologicals
NBP1-41190
102169-994EA 635.51 USD
102169-994
Anti-DNA Ligase III Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
The DNA Ligase III Antibody (1F3) from Novus Biologicals is a mouse monoclonal antibody to DNA Ligase III. This antibody reacts with human, mouse, chicken. The DNA Ligase III Antibody (1F3) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Blocking / Neutralizing, Proximity Ligation Assay.

Type: Primary
Antigen: DNA Ligase III
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR