1-Chloro-4-ethynylbenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne
066831-5G
066831-25G
101780-840EA
55.38
USD
101780-840
102833-416
1-Chloro-4-ethynylbenzene ≥95%
1-Chloro-4-ethynylbenzene
Hazard: Flammable Solid
Formula:
C₈H₅Cl Boiling Pt: 179 °C Melting Pt: 43…47 °C Flash Pt: 57 °C |
MDL Number:
MFCD00191917 CAS Number: 873-73-4 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...