2-Piperazinone ≥97%

Supplier: Matrix Scientific

Synonyms: Piperazin-2-one

012421-5G 012421-25G
101780-102EA 30.77 USD
101780-102 101780-104
2-Piperazinone ≥97%
2-Piperazinone

Hazard: Corrosive

Formula: C₄H₈N₂O
MW: 100.12 g/mol
Boiling Pt: 165 °C (5 Torr)
Melting Pt: 135…137 °C
MDL Number: MFCD01318687
CAS Number: 5625-67-2

Order Now

Specification Test Results

Melting point 135-137°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR