3-Hydroxythiophenol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxybenzene-1-thiol, 3-Hydroxybenzenethiol, 3-Hydroxythiophenol, 1-Hydroxy-3-mercaptobenzene, 3-Mercaptophenol, m-Hydroxythiophenol, 3-Hydroxyphenyl mercaptan

011292-5G 011292-25G
101780-088EA 41.54 USD
101780-088 101780-090
3-Hydroxythiophenol ≥98%
3-Hydroxythiophenol

Hazard: Corrosive/Air Sensitive/Stench

Formula: C₆H₆OS
MW: 126.18
Boiling Pt: 168 °C (15 Torr)
Melting Pt: 16.5
Density: 1.237
MDL Number: MFCD01317829
CAS Number: 40248-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR