1,1,1,2,2-Pentafluoro-3-oxopentane ≥97%

Supplier: Matrix Scientific

Synonyms: Pentafluoroethyl ethyl ketone

009077-2.5G 009077-500MG
101779-866EA 361.01 USD
101779-866 101779-864
1,1,1,2,2-Pentafluoro-3-oxopentane ≥97%
1,1,1,2,2-Pentafluoro-3-oxopentane

Hazard: Flammable

Formula: C₅H₅F₅O
MW: 176.09 g/mol
Boiling Pt: 62 °C
Density: 1.554
MDL Number: MFCD00053172
CAS Number: 378-72-3

Order Now

Specification Test Results

Boiling point 62°
Density 1.554
ND20 1.3905


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR