1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98%

Supplier: Matrix Scientific

Synonyms: Perfluoropropoxyethylene, (Heptafluoropropoxy)trifluoroethene, Perfluoro(propyl vnyl ether)

006137-5G 006137-100G 006137-25G
101778-898EA 27.69 USD
101778-898 101778-902 101778-900
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98%
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane

TCA listed: T
Hazard: Flammable/Refrigerate

Formula: C₅F₁₀O
Boiling Pt: 35…36 °C
Density: 1.53
MDL Number: MFCD00191577
CAS Number: 1623-05-8

Order Now

Specification Test Results

Boiling point 35-36°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR