2,6-Dichloroaniline ≥97%

Supplier: Matrix Scientific

005145-100G 005145-2.5KG 005145-500G
101777-978EA 21.54 USD
101777-978 101777-982 101777-980
2,6-Dichloroaniline ≥97%
2,6-Dichloroaniline

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₆H₅Cl₂N
Melting Pt: 38…41 °C
MDL Number: MFCD00007675
CAS Number: 608-31-1
UN: 3442
ADR: 6.1,II

Order Now

Specification Test Results

Melting point 38-41°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR