Octafluoroadipic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Perfluoroadipic Acid, Octafluorohexanedioic Acid, Perfluorohexanedioic Acid

004825-5G 004825-25G 004825-100G
101777-776EA 64.62 USD
101777-776 101777-778 101777-780
Octafluoroadipic acid ≥98%
Octafluoroadipic acid

Purity: 98% CAS: 336-08-3 MDL Number: MFCD00066405 Molecular Formula: C6H2F8O4 Molecular Weight: 290,07 TCA listed: T Hazard: Corrosive

Formula: C₆H₂F₈O₄
Melting Pt: 132…134 °C
MDL Number: MFCD00066405
CAS Number: 336-08-3

Order Now

Specification Test Results

Melting point 132-134°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR