2-Bromothiophene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Thienyl bromide

004198-100G 004198-500G
101777-422EA 23.08 USD
101777-422 101777-424
2-Bromothiophene ≥98%
2-Bromothiophene

TCA listed: T
Hazard: Flammable/Toxic

Formula: C₄H₃BrS
Boiling Pt: 149…151 °C
Density: 1.7
MDL Number: MFCD00005417
CAS Number: 1003-09-4
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 149-151°
Density 1.7
ND20 1.586


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR