Thiazole ≥99%

Supplier: Matrix Scientific

001011-100G 001011-25G
101775-144EA 229.23 USD
101775-144 101775-142
Thiazole ≥99%
Thiazole

TCA listed: T
Hazard: Flammable

Formula: C₃H₃NS
Boiling Pt: 117…118 °C
Density: 1.2
MDL Number: MFCD00005315
CAS Number: 288-47-1

Order Now

Specification Test Results

Boiling point 117-118°
Density 1.2
ND20 1.539


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR