2,3,6-Trifluoroaniline ≥99%
Supplier: Matrix Scientific
003613-1G
003613-5G
101776-886EA
40
USD
101776-886
101776-888
2,3,6-Trifluoroaniline ≥99%
2,3,6-Trifluoroaniline
Hazard: Toxic
Formula:
C₆H₄F₃N Boiling Pt: 150…151 °C Density: 1.39 |
MDL Number:
MFCD00012369 CAS Number: 67815-56-9 UN: 2941 ADR: 6.1,III |
Specification Test Results
Density | 1.39 |
ND20 | 1.487 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...