α,α,α-Trifluoroacetophenone ≥99%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

003604-25G 003604-100G
101776-866EA 146.15 USD
101776-866 101776-868
α,α,α-Trifluoroacetophenone ≥99%
α,α,α-Trifluoroacetophenone

TCA listed: T
Hazard: Flammable/Refrigerate/Lachrymator

Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 165
Melting Pt: -40
Density: 1.24
Flash Pt: 42 °C
MDL Number: MFCD00000420
CAS Number: 434-45-7
UN: 1224
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69025716b957e6a38ed2ae6031ab9b0f10246bd1&vid=60781a8070ecc9020bf7794534c2b3fe9acf3ebf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Boiling point 165-166°
Density 1.24
ND20 1.4595


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR