3-Chloro-α,α,α-trifluorotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Chloro-3-(trifluoromethyl)benzene, 3-Chlorobenzotrifluoride
003312-25G
003312-100G
101776-610EA
23.08
USD
101776-610
101776-612
3-Chloro-α,α,α-trifluorotoluene ≥98%
3-Chloro-α,α,α-trifluorotoluene
TCA listed: T
Hazard: Flammable
Formula:
C₇H₄ClF₃ Boiling Pt: 137…138 °C Melting Pt: –55.4 °C Density: 1.336 |
MDL Number:
MFCD00000594 CAS Number: 98-15-7 UN: 2234 ADR: 3,III |
Specification Test Results
Boiling point | 137-138° |
Density | 1.336 |
ND20 | 1.446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...