Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H,2H,2H-Perfluorodecyltriethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane, Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

002221-25G 002221-5G
101775-874EA 235.38 USD
101775-874 101775-872
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane

Purity: 97% CAS: 101947-16-4 MDL Number: MFCD00042334 Molecular Formula: C16H19F17O3Si Molecular Weight: 610,39 Boiling Point: 103-106°C/3mm Hazard: Corrosive

Formula: C₁₆H₁₉F₁₇O₃Si
MW: 610.38 g/mol
Boiling Pt: 103…106 °C (3 mmHg)
Density: 1.407
MDL Number: MFCD00042334
CAS Number: 101947-16-4

Order Now

Specification Test Results

Boiling point 103-106°/3
Density 1.407
ND20 1.342


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR