Deuterium oxide (99.9% D), MagniSolv™ for NMR spectroscopy , pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Water-D2, Heavy water
M-Clarity™ quality level = MQ200
1.13366.0025
1.13366.0100
1.13366.0500
EM1.13366.0009EA
111
USD
EM1.13366.0009
EM1.13366.0010
EM1.13366.0025
EM1.13366.0100
EM1.13366.0500
Deuterium oxide (99.9% D), MagniSolv™ for NMR spectroscopy , pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Deuterium oxide
MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.
Formula:
D₂O MW: 20 g/mol Boiling Pt: 101.3 °C (1013 hPa) Melting Pt: 3.8 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044636 CAS Number: 7789-20-0 EINECS: 232-148-9 Merck Index: 12,02984 |
Specification Test Results
Deuteration degree | ≥ 99.90 % |
Performance test (NMR-spectrum) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...