Dextran sulfate sodium salt light, powder
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran
DE134-5KGBL
DE134-100GM
DE134-25GM
700013-158EA
10318.1
USD
700013-158
700013-152
700013-156
Dextran sulfate sodium salt light, powder
Dextran sulphate sodium salt
Dextran Sulfate Sodium Salt, Powder, Light is a sodium salt of the sulfuric acid ester of Dextran. It functions as a nonsurfactant - suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Ambient |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...