Borneol ≥70.0% (contains ca. 20% Isoborneol)
Supplier: TCI America
B0525-500G
B0525-25G
TCB0525-500GEA
184.2
USD
TCB0525-500G
TCB0525-025G
Borneol ≥70.0% (contains ca. 20% Isoborneol)
Borneol
CAS Number: 507-70-0
MDL Number: MFCD00066426
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method:
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Melting Pt: 201 °C Flash Pt: 66 °C |
MDL Number:
MFCD00066426 CAS Number: 507-70-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...