Tetrafluorotetracyanoquinodimethane ≥98.0% (by total nitrogen basis), purified by sublimation

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile

T1131-100MG T1131-1G
TCT1131-100MGEA 245 USD
TCT1131-100MG TCT1131-1G
Tetrafluorotetracyanoquinodimethane ≥98.0% (by total nitrogen basis), purified by sublimation
Tetrafluorotetracyanoquinodimethane

CAS Number: 29261-33-4
MDL Number: MFCD00042382
Molecular Formula: C12F4N4
Molecular Weight: 276.15
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: Yellow
Melting point (°C): 300


Product Categories: Materials Chemistry, Organic Transistor Materials, Molecular Conductors, Highly Purified Reagents
Other Product Names / Synonyms: TCNQF4

Formula: C₁₂F₄N₄
MW: 276.15 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00042382
CAS Number: 29261-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54a242ea6965dd0cdfe759e055c23af7d6094693&vid=f38b0a32ca1e64d032819e8bfe7cecbbd170ce32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR