Streptomycin sulfate ≥95.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
S0585-500G
S0585-25G
TCS0585-500GEA
222
USD
TCS0585-500G
TCS0585-25G
Streptomycin sulfate ≥95.0% (by total nitrogen and titration analysis)
Streptomycin sulphate
CAS Number: 3810-74-0
MDL Number: MFCD00037023
Molecular Formula: C21H39N7O12
Molecular Weight: 728.69
Purity/Analysis Method:
Formula:
C₂₁H₃₉N₇O₁₂·1.5H₂SO₄ MW: 1457.4 g/mol Melting Pt: 250 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037023 CAS Number: 3810-74-0 EINECS: 223-286-0 UN: 2811 ADR: 6.1,III Merck Index: 12,08983 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...