Simvastatin ≥97.0%
Supplier: TCI America
Synonyms:
(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
S0509-100MG
S0509-1G
TCS0509-100MGEA
67.64
USD
TCS0509-100MG
TCS0509-1G
Simvastatin ≥97.0%
Simvastatin
CAS Number: 79902-63-9
MDL Number: MFCD00072007
Molecular Formula: C25H38O5
Molecular Weight: 418.57
Purity/Analysis Method:
Formula:
C25H38O5 Melting Pt: 134 °C |
MDL Number:
MFCD00072007 CAS Number: 79902-63-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...