ɑ-Sexithiophene, purified by sublimation

Supplier: TCI America

Synonyms: 6T

S0504-1G S0504-100MG
TCS0504-1GEA 142 USD
TCS0504-1G TCS0504-100MG
ɑ-Sexithiophene, purified by sublimation
alpha-Sexithiophene

CAS Number: 88493-55-4
Molecular Formula: C24H14S6
Molecular Weight: 494.74
Form: Crystal
Color: Yellow
Melting point (°C): 304


Other Product Names / Synonyms: 6T

Formula: C₂₄H₁₄S₆
MW: 494.74 g/mol
Melting Pt: 304 °C
CAS Number: 88493-55-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c6c78ef4780ccbfd0bf2495b78eacefa01cbdca&vid=fdafc97b931600055e399c1d869a0775001b4b60&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR