Artesunate ≥98.0%

Supplier: TCI America

Synonyms: 4-Oxo-4-(((3R,5aS,6R,8aS,9R,10R,12R,12aR)-3,6,9-trimethyldecahydro-3H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10-yl)oxy)butanoic acid

A2191-25G A2191-5G
TCA2191-25GEA 202 USD
TCA2191-25G TCA2191-5G
Artesunate ≥98.0%
Artesunate

CAS Number: 88495-63-0 MDL Number: MFCD00866204 Molecular Formula: C19H28O8 Molecular Weight: 384.43 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 133 Specific rotation [a]20/D: 5.5 deg (C=1, CH2Cl2) Storage Temperature: 0-10°C

Formula: C₁₉H₂₈O₈
MW: 384.43 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00866204
CAS Number: 88495-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60080b05fd8f3c88b1151ed60c100ab2bfbfe0c6&vid=60080b05fd8f3c88b1151ed60c100ab2bfbfe0c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR