Phloridzin ≥97.0%

Supplier: TCI America

Synonyms: Phlorizin hydrate

P0248-1G P0248-5G
TCP0248-001GEA 133 USD
TCP0248-001G TCP0248-5G
Phloridzin ≥97.0%
Phloridzin

CAS Number: 60-81-1 MDL Number: MFCD00006591 Molecular Formula: C21H24O10 Molecular Weight: 436.41 Purity/Analysis Method: 97.0% (T) Form: Crystal Specific rotation [a]20/D: -54 deg (C=3.2, EtOH(95vol%))

Formula: C21H24O10
MW: 436.41 g/mol
MDL Number: MFCD00006591
CAS Number: 60-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f85fb005ea30fdaa861ee067cb99c4b6724ceaaa&vid=f85fb005ea30fdaa861ee067cb99c4b6724ceaaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR