Trimethyl orthobutyrate ≥96.0%

Supplier: TCI America

Synonyms: Trimethylorthobutyrate, 1,1,1-Trimethoxybutane, Orthobutyric Acid Trimethyl Ester

O0179-25ML O0179-5ML
TCO0179-025MLEA 50 USD
TCO0179-025ML TCO0179-5ML
Trimethyl orthobutyrate ≥96.0%
Trimethyl orthobutyrate

CAS Number: 43083-12-1 MDL Number: MFCD00008480 Molecular Formula: C7H16O3 Molecular Weight: 148.20 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 146 Flash Point (°C): 35 Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: Orthobutyric Acid Trimethyl Ester, 1,1,1-Trimethoxybutane

Formula: C₇H₁₆O₃
MW: 148.20 g/mol
Boiling Pt: 146 °C
Density: 0.93 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00008480
CAS Number: 43083-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=221150bafa56aa9490cebc43693ec9fc6eb5e05b&vid=221150bafa56aa9490cebc43693ec9fc6eb5e05b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR