2,4-Mesitylenedisulfonyl dichloride ≥98.0%
Supplier: TCI America
Synonyms:
2,4,6-Trimethyl-1,3-benzenedisulphonyl Dichloride, 2,4,6-Trimethylbenzene-1,3-disulfonyl dichloride, 2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride, 2,4,6-Trimethylbenzene-1,3-disulphonyl dichloride
CAS Number: 68985-08-0
MDL Number: MFCD00009796
Molecular Formula: C9H10Cl2O4S2
Molecular Weight: 317.20
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Condensation & Active Esterification, Nucleic Acid Synthesis
Other Product Names / Synonyms: 2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride
Formula:
C₉H₁₀Cl₂O₄S₂ MW: 317.20 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00009796 CAS Number: 68985-08-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...