Monocaprylin ≥98.0%
Supplier: TCI America
CAS Number: 26402-26-6
MDL Number: MFCD00056652
Molecular Formula: C11H22O4
Molecular Weight: 218.29
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha-Monooctanoate, 1-Monooctanoyl Glycerol
Formula:
C₁₁H₂₂O₄ Melting Pt: 41 °C |
MDL Number:
MFCD00056652 CAS Number: 26402-26-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...